Learn More
NKp44 (CD336) Mouse anti-Human, BB515, Clone: p44-8, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566025
Catalog No. BDB566025
Add to cart
Description
Description
NKp44 (CD336) Monoclonal antibody specifically detects NKp44 (CD336) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
NKp44 (CD336)
p44-8
Aqueous buffered solution containing ≤0.09% sodium azide.
NCR2; NCTR2; NKp44; NK-p44; LY95; dJ149M18.1; p44-8
Human NKp44 Recombinant Protein
25 Tests
Cell Biology
Primary
Human
Flow Cytometry
BB515
Purified
Mouse
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.