Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp46/NCR1 Mouse anti-Human, Janelia Fluor 646, Clone: n1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104311JF646

Catalog No. NBP14311646


Add to cart

Description

Description

NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

NKp46/NCR1
n1D9
Flow Cytometry
Purified
NCR1
Recombinant human NKp46 protein
Protein G purified
Innate Immunity
Primary
9437
Human
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-NKp46, clone n1D9, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with recombinant human NKp46.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit