Learn More
NKp46/NCR1 Rat anti-Mouse, Alexa Fluor™ 700, Clone: 29A1.4, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB22252N100UG
Catalog No. FB22252N100
Add to cart
This item is not returnable. View return policy
Description
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
NKp46/NCR1
29A1.4
Flow Cytometry 0.25-1 μg/106 cells
Q8C567
CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Rat
IgG2a
100 μg
Primary
9437
Mouse
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant, Conjugated
NCR1
Murine myeloma cell line NSO-derived recombinant mouse NKp46-Fc chimera , Glu22-Asn255, Accession # Q8C567
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse NKp46/NCR1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp44, rhNKp46, or rhNKp80 is observed.