Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKp80 Mouse anti-Human, Clone: 5D12, MilliporeSigma™
Mouse Monoclonal Antibody
$190.00 - $450.00
Specifications
Antigen | NKp80 |
---|---|
Clone | 5D12 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Immunoprecipitation, Inhibition Assays |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2223100
|
MilliporeSigma
MABF2223100UG |
100 μg |
Each for $450.00
|
|
|||||
MABF222325
|
MilliporeSigma
MABF222325UG |
25 μL |
Each for $190.00
|
|
|||||
Description
NKp80 Monoclonal antibody specifically detects NKp80 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Inhibition.Specifications
NKp80 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing PBS without preservatives. | |
IgG1 κ | |
Protein G purified |
5D12 | |
Flow Cytometry, Immunoprecipitation, Inhibition Assays | |
Unconjugated | |
Mouse | |
Human | |
Recombinant extracellular domain of human NKp80. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title