Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKRF Antibody (OTI4C2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273001AF405
Description
NKRF Monoclonal specifically detects NKRF in Human, Mouse samples. It is validated for Western Blot.Specifications
| NKRF | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NKRF | |
| Recombinant protein fragment corresponding to amino acids 1-296 of human NKRF (NP_060014) produced in E.coli. | |
| 0.1 mL | |
| 55922.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4C2 | |
| Western Blot | |
| ITBA4NFkB-repressing factor, NF-kappaB repressing factor, NFKB repressing factor, NRFNF-kappa-B-repressing factor, Protein ITBA4, Transcription factor NRF | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction