Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nkx2.3 Polyclonal Antibody
Nkx2.3 Polyclonal Antibody
Supplier: Thermo Scientific PA598740
Description
Nkx2.3 Polyclonal Antibody for Western Blot, ELISA

Specifications
Nkx2.3 | |
Polyclonal | |
Unconjugated | |
NKX2-3 | |
CSX3; Drosophila NK2 transcription factor related, locus 3; homeobox protein NK-2 homolog 3; Homeobox protein NK-2 homolog C; homeobox protein Nkx-2.3; homeobox protein Nkx-2.3; LOW QUALITY PROTEIN: homeobox protein Nkx-2.3; hypothetical protein LOC539451; NK2 homeobox 3; NK2 transcription factor related, locus 3; NK2.3; NKX2.3; Nkx-2.3; NKX23; NKX2-3; NKX2C; Nkx2-C; NKX4-3; tinman | |
Rabbit | |
Protein G | |
RUO | |
159296 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.76 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8TAU0 | |
NKX2-3 | |
Recombinant Human Homeobox protein Nkx-2.3 protein (52-148AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction