Learn More
Nkx2.5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549431
Catalog No. PA549431
Add to cart
Description
Description
NKX2-5 is a transcription factor protein which plays a crucial role in cardiac development and differentiation. It is a low molecular weight protein of 35-38 kDa depending on phosphorylation status. Expression is mainly restricted to the heart. But during embryogenesis NKX2-5 expression has also been detected in spleen-precursor cells. Over expression of NKX2-5 resulted in expression of neuronal markers in C2C12 murine myogenic cells. Functionally it is involved in differentiation processes in heart development and in homeostasis and survival of cardiac myocytes as well maintains the initial muscle differentiation paradigm.
Specifications
Specifications
Nkx2.5
Polyclonal
Unconjugated
NKX2-5
Liquid
NKX2-5
KLH conjugated synthetic peptide between 98-133 amino acids from the Central region of mouse Nkx2-5.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P52952, P42582
Homeobox protein Nkx-25, Cardiac-specific homeobox, Homeobox protein CSX, Homeobox protein NK-2 homolog E, Nkx2-5, Csx, Nkx-25, Nkx2e
Rabbit
IgG
200μl
-20° C, Avoid Freeze/Thaw Cycles
Primary
1482, 18091