Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2.8 Mouse anti-Human, Clone: NKX28/2547, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30761110UG
Description
NKX2.8 Monoclonal specifically detects NKX2.8 in Human samples. It is validated for Western Blot.Specifications
NKX2.8 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 0.5 ug-1 ug/ml | |
Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H | |
Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
NKX28/2547 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
26257 | |
Store at 4°C. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title