Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.8 Mouse anti-Human, Clone: NKX28/2547, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30761120UG

 View more versions of this product

Catalog No. NB122696


Add to cart

Description

Description

NKX2.8 Monoclonal antibody specifically detects NKX2.8 in Human samples. It is validated for Western Blot
Specifications

Specifications

NKX2.8
NKX28/2547
Unconjugated
10 mM PBS with 0.05% BSA
Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H
Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
26257
Western Blot
0.2 mg/ml
Western Blot 0.5 ug-1 ug/ml
Purified
Mouse
IgG1 Kappa
20 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit