Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.8 Mouse anti-Human, Clone: NKX28/2547, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen NKX2.8
Clone NKX28/2547
Concentration 0.2 mg/ml
Dilution Western Blot 0.5 ug-1 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122696
View Documents
Novus Biologicals
NBP30761120UG
20 μg
Each for $138.37
Add to cart
 
NB122695
View Documents
Novus Biologicals
NBP30761110UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

NKX2.8 Monoclonal antibody specifically detects NKX2.8 in Human samples. It is validated for Western Blot
Specifications

Specifications

NKX2.8
0.2 mg/ml
Western Blot
Purified
RUO
10 mM PBS with 0.05% BSA
26257
IgG1 Kappa
Primary
Store at 4°C.
NKX28/2547
Western Blot 0.5 ug-1 ug/ml
Unconjugated
Mouse
Human
Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H
Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit