Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX2.8 Rabbit anti-Human, Clone: NKX28/3233R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | NKX2.8 |
---|---|
Clone | NKX28/3233R |
Concentration | 0.2 mg/ml |
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122748
|
Novus Biologicals
NBP30763720UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
NKX2.8 Monoclonal specifically detects NKX2.8 in Human samples. It is validated for Western Blot, ELISA.Specifications
NKX2.8 | |
0.2 mg/ml | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H | |
Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
NKX28/3233R | |
Western Blot, ELISA | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
26257 | |
IgG | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title