Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX3.1, Mouse anti-Human, Alexa Fluor 750, Clone: NKX3.1/2576, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279871AF750
Description
NKX3.1 Monoclonal antibody specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
NKX3.1 | |
NKX3.1/2576 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
NKX3-1 | |
Recombinant fragment (around aa 92-224) of human NKX3.1 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
Mouse | |
IgG2c Kappa | |
0.1 mL | |
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title