Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX3.1 Mouse anti-Human, Clone: NKX3.1/2836, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30743510UG
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
NKX3.1 | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
Recombinant fragment (around aa 92-224) of human NKX3.1 protein (exact sequence is proprietary) | |
100 μg | |
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
4824 | |
Store at 4°C. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
NKX3.1/2836 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title