Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3.1 Mouse anti-Human, Clone: NKX3.1/3348, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen NKX3.1
Clone NKX3.1/3348
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122347
View Documents
Novus Biologicals
NBP30743710UG
100 μg
Each for $352.89
Add to cart
 
NB122348
View Documents
Novus Biologicals
NBP30743720UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

NKX3.1 Monoclonal antibody specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

NKX3.1
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A
Recombinant fragment(around aa 92-224) of humanNKX3.1 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
NKX3.1/3348
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators
10 mM PBS with 0.05% BSA
4824
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit