Learn More
Nkx3.1 Mouse anti-Human, Clone: OTI1C11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805087
Catalog No. 50-166-7274
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Specifications
Specifications
Nkx3.1
Monoclonal
Unconjugated
NKX3-1
Lyophilized
NKX3-1
Full length human recombit protein of human NKX3-1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1C11
PBS with 8% trehalose and no preservative; pH 7.3
Q99801
BAPX2, NKX3, NKX3.1, NKX3A
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4824