Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKX6.1, Mouse anti-Human, Janelia Fluor 646, Clone: NKX61/2561, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279872JF646
Description
NKX6.1 Monoclonal antibody specifically detects NKX6.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
NKX6.1 | |
NKX61/2561 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
NKX6-1 | |
Recombinant full-length human NKX6.1 protein | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1 | |
Mouse | |
IgG2c Kappa | |
0.1 mL | |
Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title