Learn More
[Nle13, Glu14] Motilin, Human, Porcine, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66270MCR
 View more versions of this product
Catalog No. AAJ66270MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2682.10
Human, Ovine, Rat
Solid
RUO
[Nle13, Glu14] Motilin, human, porcine
HPLC
1mg
Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Nle-Glu-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
-30° to -10°C