Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLE1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12401R

Description
NLE1 plays a role in regulating Notch activity, in regulating the expression of CDKN1A and several members of the Wnt pathway, probably via its effects on Notch activity. NLE1 is required during embryogenesis for inner mass cell survival.Specifications
NLE1 | |
Polyclonal | |
Unconjugated | |
NLE1 | |
AL022765; BC018399; FLJ10458; HUSSY-07; l11Jus1; l11Jus4; Nle; NLE1; Notchless gene homolog; notchless homolog 1; notchless homolog 1 (Drosophila); Notchless protein homolog 1; novel WD40 repeat containing protein; RGD1308301 | |
Rabbit | |
Protein A | |
RUO | |
54475 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NVX2 | |
NLE1 | |
KLH conjugated synthetic peptide derived from human NLE1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction