Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLGN3 Mouse anti-Human, Mouse, Rat, Clone: S110-29, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527621

Catalog No. PIMA527621


Add to cart

Description

Description

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

NLGN3
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q62889, Q8BYM5, Q9NZ94
Gliotactin homolog, Neuroligin-3, Nlgn3, NLGN3_HUMAN
Mouse
IgG1
100 μg
-20°C
Primary
171297, 245537, 54413
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S110-29
Unconjugated
NLGN3
Liquid
NLGN3
Fusion protein amino acids 730-848 (intracellular C-terminus) of rat Neuroligin-3. Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ∽60% identity with Neuroligin-1. ∽40% identity with Neuroligin-2.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit