Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nlgn4l Mouse anti-Mouse, HRP, Clone:S98-7, Abnova™

Mouse monoclonal antibody raised against partial recombinant mouse Nlgn4l.

Manufacturer:  Abnova Corporation MAB17094

Catalog No. 89-333-718


Add to cart

Specifications

Specifications

Nlgn4l
S98-7
neuroligin 4-like
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Liquid
Mouse
IgG1
Store at -20°C.
Monoclonal
Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
HRP
Immunocytochemistry
Immunofluorescence
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Nlgn4l
NL-4, NL4, Nlgn4, Nlgn4x
Recombinant protein corresponding to amino acids 782-945 at C-terminus of mouse Nlgn4l.
100μg
Antibody
100113365
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit