Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nlgn4l, Mouse anti-Mouse, PE/ATTO594, Clone: S98-7, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18676

Catalog No. 89-338-601


Add to cart

Description

Description

Specifications

Specifications

Nlgn4l
S98-7
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
neuroligin 4-like
NL-4|NL4|Nlgn4|Nlgn4x
Mouse
IgG1
RUO
Antibody
Monoclonal
Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
PE/ATTO594
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Nlgn4l
Recombinant protein corresponding to amino acids 782-945 at C-terminus of mouse Nlgn4l.
100 μg
Store at -20°C.
Primary
100113365
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit