Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLN Mouse anti-Human, Mouse, Rat, Clone: OTI2H1, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504180

Catalog No. 50-167-4925


Add to cart

Description

Description

Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides.
Specifications

Specifications

NLN
Monoclonal
0.94 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42676, Q91YP2, Q9BYT8
AGTBP, EP24.16, MEP, MOP
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
117041, 57486, 75805
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2H1
Unconjugated
NLN
Liquid
NLN
Full length human recombit protein of human NLN produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit