Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NLRC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527686
Description
Recommended positive controls: MCF-7, human NLRC4-transfected 293T. Predicted reactivity: Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NLRC4 encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
Specifications
NLRC4 | |
Polyclonal | |
Unconjugated | |
Nlrc4 | |
9530011P19Rik; AIFEC; CARD, LRR, and NACHT-containing protein; Card12; caspase recruitment domain family, member 12; caspase recruitment domain-containing protein 12; CLAN; Clan protein; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; ice protease-activating factor; ICE-protease activating factor; ipaf; NLR family CARD domain containing 4; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NLRC4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NPP4 | |
Nlrc4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 168 of Human CARD12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction