Learn More
Medchemexpress LLC NLRP3-IN-11 | 2769040-91-5 | 344.8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY148187100MG
Encompass_Preferred
NLRP3-IN-11 is a NLR family pyrin domain containing 3 (NLRP3) proteins inhibitor with an IC50 value of <0.3 μM for NLRP3. It is used in the research of inflammatory and degenerative diseases including NASH, atherosclerosis, other cardiovascular diseases, Alzheimer's disease, Parkinson's disease, diabetes, and gout. This product should be stored at 4°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
Catalog No. 50-003-63777