Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP92 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZFP92 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZFP92 Polyclonal specifically detects ZFP92 in Human samples. It is validated for Western Blot.Specifications
ZFP92 | |
Polyclonal | |
Rabbit | |
Human | |
zfp-92, zinc finger protein 92 homolog, zinc finger protein 92 homolog (mouse), zinc finger protein homologous to Zfp92 in mouse, ZNF897 | |
ZFP92 | |
IgG | |
44 kDa |
Western Blot | |
Unconjugated | |
RUO | |
XP_066859 | |
139735 | |
Synthetic peptide directed towards the middle region of human ZFP92. Peptide sequence IGDRTQSKTSTSTQKHSGRQLPGADPQGGKEGQAARSSVLQRGAQGLGQS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title