Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | NAPB |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19151520
![]() |
Novus Biologicals
NBP19151520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191515
![]() |
Novus Biologicals
NBP191515 |
100 μL |
Each for $487.50
|
|
|||||
Description
NAPB Polyclonal specifically detects NAPB in Human samples. It is validated for Western Blot.Specifications
NAPB | |
Polyclonal | |
Rabbit | |
NP_071363 | |
63908 | |
Synthetic peptide directed towards the N terminal of human NAPB. Peptide sequence MDNAGKEREAVQLMAEAEKRVKASHSFLRGLFGGNTRIEEACEMYTRAAN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC26066, N-ethylmaleimide-sensitive factor attachment protein, beta, SNAP-BETA | |
NAPB | |
IgG | |
33 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title