Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTGR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | MTGR1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MTGR1 Polyclonal specifically detects MTGR1 in Mouse samples. It is validated for Western Blot.Specifications
MTGR1 | |
Polyclonal | |
Rabbit | |
NP_766448 | |
9139 | |
Synthetic peptide directed towards the middle region of mouse CBFA2T2H. Peptide sequence RRSMAVLRRCQESDREELNYWKRRFNENTELRKTGTELVSRQHSPGSTDS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
core-binding factor, runt domain, alpha subunit 2; translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3 | |
CBFA2T2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title