Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNPTAB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$501.50
Specifications
Antigen | GNPTAB |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GNPTAB Polyclonal specifically detects GNPTAB in Human samples. It is validated for Western Blot.Specifications
GNPTAB | |
Polyclonal | |
Rabbit | |
Q3T906 | |
79158 | |
Synthetic peptide directed towards the N terminal of human GNPTAB (NP_077288). Peptide sequence FQFGEVVLEWSRDQYHVLFDSYRDNIAGKSFQNRLCLPMPIDVVYTWVNG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp762B226, GlcNAc phosphotransferase, glucosamine (UDP-N-acetyl)-lysosomal-enzyme N-acetylglucosamine phosphotransferase, GNPTA, ICD, KIAA1208, MGC4170, N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits, stealth protein GNPTAB, UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosamine | |
GNPTAB | |
IgG | |
144 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title