Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NM23-H1 Mouse anti-Human, Mouse, Rat, Clone: OTI4H2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273003
Description
NM23-H1 Monoclonal specifically detects NM23-H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NM23-H1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein | |
Mouse | |
19.5 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI4H2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
NME1 | |
Full length human recombinant protein of human NME1 (NP_937818) produced in E.coli. | |
Affinity Purified | |
Lipid and Metabolism | |
4830 | |
Human, Mouse, Rat | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title