Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NM23-H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen NM23-H1
Applications Immunofluorescence, Immunohistochemistry, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092422
View Documents
Novus Biologicals
NBP303013100UL
100 μg Each for $320.71
Add to cart
 
NB092423
View Documents
Novus Biologicals
NBP30301320UL
20 μg Each for $138.37
Add to cart
 
Description

Description

NM23-H1 Polyclonal antibody specifically detects NM23-H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Specifications

Specifications

NM23-H1
Unconjugated
Rabbit
Lipid and Metabolism
PBS with 50% glycerol, pH7.3.
4830
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein
A synthetic peptide corresponding to a sequence within amino acids 90 to the C-terminus of human NM23-H1 (NP_000260.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit