Learn More
NMD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160601AP
Catalog No. 50-173-0684
Add to cart
Description
Description
NMD3 is a cytoplasmic factor required for a late cytoplasmic assembly step of the 60S subunit but is not a ribosomal protein. A mutation in NMD3 was found to be lethal in the absence of XRN1, which encodes the major cytoplasmic exoribonuclease responsible for mRNA turnover. The NMD3 protein sequence does not contain readily recognizable motifs of known function. However, NMD3 orthologues display an amino-terminal domain containing four repeats of Cx2C, reminiscent of zinc-binding proteins, implicated in nucleic acid binding or protein oligomerization.
Specifications
Specifications
NMD3
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96D46, Q99L48
CGI 07, hNMD3, NMD3
Rabbit
IgG
150 μL
-20°C
Primary
310512, 51068, 97112
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
NMD3
Liquid
NMD3
NMD3 Fusion Protein Ag8894
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat