Learn More
NMDAR1 Mouse anti-Human, Mouse, Rat, HRP, Clone: S308-48, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259327H
Catalog No. NBP259327H
Add to cart
Description
Description
NMDAR1 Monoclonal antibody specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
NMDAR1
S308-48
Western Blot, Immunohistochemistry
Purified
GRIN1
Fusion protein amino acids 42-361 (extracellular N-terminus) of rat NR1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 105kDa.
Western Blot, Immunohistochemistry
HRP
PBS with No Preservative
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1
Mouse
IgG1
100 ul
Primary
2902
Human, Mouse, Rat