Learn More
NMDAR2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 199531AP
Catalog No. 50-173-2620
Add to cart
Description
Description
The antibody recognize the C-term of GRIN2A.
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
NMDAR2A
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35436, Q12879
GluN2A, GRIN2A, hNR2A, NMDAR2A, NR2A
Rabbit
IgG
150 μL
-20°C
Primary
14811, 2903
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
GRIN2A
Liquid
Grin2a
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse