Learn More
NMDAR2B Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: S59-36, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147611C
Catalog No. NBP147611C
Add to cart
Description
Description
NMDAR2B Monoclonal antibody specifically detects NMDAR2B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
NMDAR2B
S59-36
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q00960
glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 166kDa. No cross-reactivity against NR2A.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Purified
GRIN2B
Fusion protein amino acids 20-271 (extracellular N-terminus) of rat NR2B
Protein G purified
Autophagy, Neurodegeneration, Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary
2904
Human, Mouse, Rat