Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2D Mouse anti-Rat, Clone: 1G9.39A5, MilliporeSigma™

Mouse Monoclonal Antibody

$143.00 - $365.00

Specifications

Antigen NMDAR2D
Clone 1G9.39A5
Concentration 1.0 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5578I100
View Documents
MilliporeSigma
MAB5578I100UG
100 μg
Each for $365.00
Add to cart
 
MAB5578I25
View Documents
MilliporeSigma
MAB5578I25UG
25 μg
Each for $143.00
Add to cart
 
Description

Description

NMDAR2D Monoclonal antibody specifically detects NMDAR2D in Rat samples. It is validated for Western Blotting.
Specifications

Specifications

NMDAR2D
1.0 mg/mL
Unconjugated
Mouse
Rat
Recombinant rat NMDAR2D.
Monoclonal
Protein G purified
1G9.39A5
Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2b
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit