Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDARA1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281973PCP
Description
NMDARA1 Polyclonal antibody specifically detects NMDARA1 in Human samples. It is validated for Western Blot, ELISASpecifications
| NMDARA1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| glutamate [NMDA] receptor-associated protein 1, glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1(glutamate binding), glutamate receptor, NMDA subtype, glutamate-binding subunit, HNRGW, LFG1, NMDA receptor glutamate-binding subunit, NMDARA1MGC99687, Putative MAPK-activating protein PM02, TMBIM3, transmembrane BAX inhibitor motif containing 3 | |
| Rabbit polyclonal NMDARA1 antibody was raised against an 18 amino acid peptide near the center of human NMDARA1. The immunogen is located within amino acids 110 - 160 of NMDARA1. Amino Acid Squence: QDPDSPQHGNYQEEGPPS | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 2907 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction