Learn More
NME3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151361AP
Catalog No. 50-173-0011
Add to cart
Description
Description
NME3 mRNA is preferentially expressed at early stages of myeloid differentiation of highly purified CD34(+) cells. Its constitutive expression in a myeloid precursor line, which is growth-factor dependent for both proliferation and differentiation, results in inhibition of granulocytic differentiation induced by granulocyte colony-stimulating factor and causes apoptotic cell death. These results appear consistent with a role for the NME3 gene in normal hematopoiesis and raise the possibility that its overexpression contributes to differentiation arrest, a feature of blastic transformation in chronic myelogenous leukemia.
Specifications
Specifications
NME3
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13232, Q9WV85
c371H6.2, DR nm23, KIAA0516, NDK 3, NDP kinase 3, NDPK C, NDPKC, NME3
Rabbit
IgG
150 μL
-20°C
Primary
4832, 79059, 85269
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
NME3
Liquid
NME3
NME3 Fusion Protein Ag7284
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat