Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NME6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550245
Description
The antibody detects endogenous levels of total NME6 protein.
NME6 Polyclonal antibody specifically detects NME6 in Human, Mouse samples. It is validated for Western Blot.Specifications
NME6 | |
Polyclonal | |
Unconjugated | |
NME6 | |
expressed in non-metastatic cells 6 (nucleoside-diphosphate kinase); expressed in non-metastatic cells 6 protein (nucleoside diphosphate kinase); expressed in non-metastatic cells 6, protein (nucleoside diphosphate kinase); inhibitor of p53-induced apoptosis-alpha; IPIA-alpha; NDK 6; NDP kinase 6; nm23/nucleoside diphosphate kinase 6; nm23-H6; nm23-M6; Nm23-R6; NME/NM23 nucleoside diphosphate kinase 6; Nme6; non-metastatic cells 6, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase 6; nucleoside diphosphate kinase type 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10201, 54369 | |
-20°C | |
Liquid |
Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O75414, O88425 | |
NME6 | |
Full length fusion protein of Human NME6 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction