Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550813

Catalog No. PA550813


Add to cart

Description

Description

The antibody detects endogenous levels of total NME8 / TXNDC3 protein.

This gene encodes a protein with an N-terminal thioredoxin domain and three C-terminal nucleoside diphosphate kinase (NDK) domains, but the NDK domains are thought to be catalytically inactive. The sea urchin ortholog of this gene encodes a component of sperm outer dynein arms, and the protein is implicated in ciliary function. Mutations in this gene are implicated in primary ciliary dyskinesia type 6.
Specifications

Specifications

TXNDC3
Polyclonal
Unconjugated
NME8
Liquid
NME8
Fusion protein corresponding to residues near the C terminal of human NME/NM23 family member 8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8N427
SPTRX2, TXNDC3
Rabbit
IgG
100 μL
-20°C
Primary
51314
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit