Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMI Mouse anti-Human, Clone: 1580CT730.43.59, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524742

Catalog No. PIMA524742

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.
Specifications

Specifications

NMI
Monoclonal
0.4 mg/mL
PBS with 0.09% sodium azide
Q13287
N-myc-interactor, Nmi, N-myc and STAT interactor, NMI
Mouse
IgG1, kappa
200 μL
-20°C
Primary
9111
Western Blot
1580CT730.43.59
Unconjugated
NMI
Liquid
NMI
human NMI recombit protein
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit