Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMNAT-1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297227APC
Description
NMNAT-1 Polyclonal antibody specifically detects NMNAT-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
NMNAT-1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 2.7.7.1, EC 2.7.7.18, NaMN adenylyltransferase 1, nicotinamide mononucleotide adenylyltransferase 1, nicotinamide nucleotide adenylyltransferase, nicotinamide nucleotide adenylyltransferase 1, Nicotinate-nucleotide adenylyltransferase 1, NMN adenylyltransferase 1, NMNATPNAT1, pyridine nucleotide adenylyltransferase 1 | |
Produced in rabbits immunized with E. coli-derived Human NMNAT-1 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
64802 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction