Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMNAT2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548838

Catalog No. PA548838

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate but with a lower efficiency. Cannot use triazofurin monophosphate (TrMP) as substrate. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity prefers NAD(+), NADH and NAAD as substrates and degrades nicotinic acid adenine dinucleotide phosphate (NHD) less effectively. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+).

Predicted to react with bovine, mouse and rat based on sequence homology.
Specifications

Specifications

NMNAT2
Polyclonal
PBS with 0.09% sodium azide
Q9BZQ4
Nicotinamide mononucleotide adenylyltransferase 2, NMN adenylyltransferase 2, Nicotinate-nucleotide adenylyltransferase 2, NaMN adenylyltransferase 2, NMNAT2, C1orf15, KIAA0479
Rabbit
IgG
400μL
Store at 2° to 8°C for up to 6 months. For long term storage store at -20°C.
Primary
23057
Western Blot
Unconjugated
NMNAT2
Liquid
NMNAT2
KLH conjugated synthetic peptide between 13-40 amino acids from the N-terminal region of human NMNAT2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit