Learn More
Abnova™ NMNAT3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00349565T01
Catalog No. 89-954-317
Add to cart
Specifications
Specifications
293T
NM_178177.2
pCMV-NMNAT3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NMNAT3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 23.76kDa
Transient overexpression cell lysate was tested with anti-NMNAT3 antibody (Mfr. No. H00349565-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human