Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMP (1-Methyl-2-pyrrolidinone), BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
872-50-4
C5H9NO
99.133
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.133
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 99.13
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.03kg
Flash Point 96.00°C
Quantity 500mL
Chemical Name or Material NMP (1-Methyl-2-pyrrolidinone)