Learn More
Norell IncSupplier Diversity Partner NMR TUBES 4IN 5MM CAPS 96PK DFS Item
Manufacturer:  Norell IncSupplier Diversity Partner SVOP5103.596
5mm OD, 103.5mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES OPEN PORT SAMPLE VAULT CAPS
Catalog No. NC0105287