Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMT2 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: OTI1G3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273021FR
Description
NMT2 Monoclonal specifically detects NMT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NMT2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
NMT2 | |
Full length recombinant protein of human NMT2 (NP_004799) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1G3 | |
Western Blot, Immunohistochemistry | |
EC 2.3.1.97, glycylpeptide N-tetradecanoyltransferase 2, Myristoyl-CoA:protein N-myristoyltransferase 2, NMT 2, N-myristoyltransferase 2, Peptide N-myristoyltransferase 2, Type II N-myristoyltransferase | |
Mouse | |
Affinity Purified | |
Cancer | |
9397.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title