Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMT2 Mouse anti-Human, Mouse, Rat, Clone: OTI2A5, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504177

Catalog No. 50-167-4923


Add to cart

Description

Description

N-myristoyltransferase (NMT) catalyzes the reaction of N-terminal myristoylation of many signaling proteins. It transfers myristic acid from myristoyl coenzyme A to the amino group of a protein ft.s N-terminal glycine residue. Biochemical evidence indicates the presence of several distinct NMTs, varying in apparent molecular weight and /or subcellular distribution. The predicted 498-amino acid of human NMT2 protein shares 77% and 96% sequence identity with human NMT1 and mouse Nmt2 comprise two distinct families of N-myristoyltransferases.
Specifications

Specifications

NMT2
Monoclonal
0.79 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60551, O70311
N-myristoyltransferase 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18108, 291318, 9397
Immunohistochemistry (Paraffin), Western Blot
OTI2A5
Unconjugated
NMT2
Liquid
Nmt2
Full length human recombit protein of human NMT2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit