Learn More
NNC 05-2090 hydrochloride, Tocris
GABA uptake inhibitor; moderately BGT-1 selective
Manufacturer:  R&D Systems 2747/50
Quantity 50mg
 View more versions of this product
Catalog No. 27-475-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
451.01
NNC 05-2090 hydrochloride
C27H30N2O2·HCl
1-[3-(9H-Carbazol-9-yl)propyl]-4-(2-methoxyphenyl)-4-piperidinol hydrochloride
Specifications
50mg
>98%
451.01
NNC 05-2090 hydrochloride
184845-18-9
C27H30N2O2·HCl
1-[3-(9H-Carbazol-9-yl)propyl]-4-(2-methoxyphenyl)-4-piperidinol hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature