NNC 05-2090 hydrochloride, Tocris

GABA uptake inhibitor; moderately BGT-1 selective

Manufacturer: R&D Systems 2747/10

Quantity 10mg

 View more versions of this product

Catalog No. 27-471-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C27H30N2O2·HCl
    Formula Weight 451.01
    Synonym 1-[3-(9H-Carbazol-9-yl)propyl]-4-(2-methoxyphenyl)-4-piperidinol hydrochloride
    Chemical Name or Material NNC 05-2090 hydrochloride

    Specifications

    Formula Weight 451.01
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C27H30N2O2·HCl
    CAS 184845-18-9
    Synonym 1-[3-(9H-Carbazol-9-yl)propyl]-4-(2-methoxyphenyl)-4-piperidinol hydrochloride
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material NNC 05-2090 hydrochloride

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit