Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NNMT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548630

Catalog No. PA548630

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor. [provided by RefSeq].
Specifications

Specifications

NNMT
Polyclonal
PBS with 0.09% sodium azide
P40261
Nicotinamide N-methyltransferase, NNMT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4837
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NNMT
Liquid
NNMT
KLH conjugated synthetic peptide between 101-130 amino acids from the Central region of human NNMT.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit